Propyltriethoxysilane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Triethoxy(propyl)silane

T2867-25G T2867-100G
TCT2867-25GEA 19.29 USD
TCT2867-25G TCT2867-100G
Propyltriethoxysilane ≥98.0% (by GC)
Propyltriethoxysilane

CAS Number: 2550-02-9
MDL Number: MFCD00026756
Molecular Formula: C9H22O3Si
Molecular Weight: 206.36
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 180
Flash Point (°C): 45
Specific Gravity (20/20): 0.89


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Propyltriethoxysilane

Formula: C₉H₂₂O₃Si
MW: 206.36 g/mol
Boiling Pt: 180 °C
Density: 0.89 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00026756
CAS Number: 2550-02-9
https://us.vwr.com/stibo/web/std.lang.all/65/19/28996519.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR