4-Methylphenethylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-(p-Tolyl)ethylamine
T2864-5ML
TCT2864-5MLEA
169.51
USD
TCT2864-5ML
4-Methylphenethylamine ≥98.0% (by GC, titration analysis)
4-Methylphenethylamine
CAS Number: 3261-62-9
MDL Number: MFCD00008195
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 215
Flash Point (°C): 90
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: 2-(4-Methylphenyl)ethylamine, 4-Methylphenethylamine
Formula:
C₉H₁₃N MW: 135.21 g/mol Boiling Pt: 215 °C Density: 0.95 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00008195 CAS Number: 3261-62-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...