3,3',4,5'-Tetramethoxypiceatannol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (E)-4-(3,5-Dimethoxystyryl)-1,2-dimethoxybenzene

T2842-1G T2842-5G
TCT2842-5GEA 383.5 USD
TCT2842-5G TCT2842-1G
3,3',4,5'-Tetramethoxypiceatannol ≥98.0% (by GC)
3,3',4,5'-Tetramethoxypiceatannol

CAS Number: 83088-26-0
MDL Number: MFCD09033675
Molecular Formula: C18H20O4
Molecular Weight: 300.35
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 69


Product Categories: Bioscience, Aromatic Polyketides
Other Product Names / Synonyms: 3,3',4,5'-Tetramethoxy-trans-stilbene

Formula: C₁₈H₂₀O₄
MW: 300.3490 g/mol
Melting Pt: 69 °C
MDL Number: MFCD09033675
CAS Number: 83088-26-0
https://us.vwr.com/stibo/web/std.lang.all/65/02/28996502.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR