Tetrabenazine ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Isobutyl-9,10-dimethoxy-1,3,4,6,7,11b-hexahydro-2H-benzo[a]quinolizin-2-one

T2839-1G T2839-200MG
TCT2839-1GEA 706.23 USD
TCT2839-1G TCT2839-200MG
Tetrabenazine ≥99.0% (by HPLC, titration analysis)
Tetrabenazine

CAS Number: 58-46-8
Molecular Formula: C19H27NO3
Molecular Weight: 317.43
Purity/Analysis Method: 99.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 127
Storage Temperature: 0-10°C


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 3-Isobutyl-9,10-dimethoxy-1,3,4,6,7,11b-hexahydro-2H-benzo[a]quinolizin-2-one

Formula: C₁₉H₂₇NO₃
MW: 317.43 g/mol
Melting Pt: 127 °C
CAS Number: 58-46-8
https://us.vwr.com/stibo/web/std.lang.all/65/00/28996500.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR