3,4,5-Trimethoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(3,4,5-Trimethoxyphenyl)metaboric acid
T2837-1G
TCT2837-1GEA
128
USD
TCT2837-1G
3,4,5-Trimethoxyphenylboronic acid (contains varying amounts of Anhydride)
3,4,5-Trimethoxyphenylboronic acid
CAS Number: 182163-96-8
MDL Number: MFCD01075676
Molecular Formula: C9H13BO5
Molecular Weight: 212.01
Form: Crystal
Color: White
Melting point (°C): 247
Other Product Names / Synonyms: 3,4,5-Trimethoxybenzeneboronic Acid
Formula:
C₉H₁₃BO₅ MW: 212.01 g/mol Melting Pt: 247 °C |
MDL Number:
MFCD01075676 CAS Number: 182163-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...