1,3,5-Tris(trifluoromethyl)benzene ≥98.0% (by GC)
Supplier: TCI America
T2830-5G
TCT2830-5GEA
134.18
USD
TCT2830-5G
1,3,5-Tris(trifluoromethyl)benzene ≥98.0% (by GC)
1,3,5-Tris(trifluoromethyl)benzene
CAS Number: 729-81-7
MDL Number: MFCD00054722
Molecular Formula: C9H3F9
Molecular Weight: 282.11
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 120
Flash Point (°C): 43
Specific Gravity (20/20): 1.52
Formula:
C₉H₃F₉ Boiling Pt: 120 °C Density: 1.52 (20 °C) Flash Pt: 43 °C |
MDL Number:
MFCD00054722 CAS Number: 729-81-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...