4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl bromide ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
3-[4-(Bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine
T2819-1G
T2819-200MG
TCT2819-200MGEA
149.16
USD
TCT2819-200MG
TCT2819-1G
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl bromide ≥95.0% (by HPLC)
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl bromide
CAS Number: 92367-11-8
Molecular Formula: C9H6BrF3N2
Molecular Weight: 279.06
Purity/Analysis Method:
Form: Clear Liquid
Color: Pale Yellow
Storage Temperature: 0-10°C
Product Categories: Bioscience, Photoaffinity Labels, Crosslinkers
Other Product Names / Synonyms: 3-[4-(Bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine
Formula:
C₉H₆BrF₃N₂ MW: 279.06 g/mol |
MDL Number:
MFCD19442581 CAS Number: 92367-11-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...