Triptycene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 9,10-Dihydro-9,10-[1,2]benzenoanthracene

T2807-1G
TCT2807-1GEA 37.83 USD
TCT2807-1G
Triptycene ≥98.0% (by GC)
Triptycene

CAS Number: 477-75-8
MDL Number: MFCD00003813
Molecular Formula: C20H14
Molecular Weight: 254.33
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 255


Other Product Names / Synonyms: 9,10-Dihydro-9,10-[1,2]benzenoanthracene

Formula: C₂₀H₁₄
MW: 254.33 g/mol
Melting Pt: 255 °C
MDL Number: MFCD00003813
CAS Number: 477-75-8
https://us.vwr.com/stibo/web/std.lang.all/64/82/28996482.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR