2,3,6,7-Tetrakis(2-cyanoethylthio)tetrathiafulvalene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

T2806-1G T2806-100MG
TCT2806-1GEA 454.23 USD
TCT2806-1G TCT2806-100MG
2,3,6,7-Tetrakis(2-cyanoethylthio)tetrathiafulvalene ≥98.0% (by HPLC, total nitrogen)
2,3,6,7-Tetrakis(2-cyanoethylthio)tetrathiafulvalene

CAS Number: 132765-36-7
Molecular Formula: C18H16N4S8
Molecular Weight: 544.83
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 191

Formula: C₁₈H₁₆N₄S₈
MW: 544.83 g/mol
Melting Pt: 191 °C
CAS Number: 132765-36-7
https://us.vwr.com/stibo/web/std.lang.all/64/81/28996481.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR