Thiamphenicol ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2,2-Dichloro-N-[(1R,2R)-1,3-dihydroxy-1-[4-(methylsulfonyl)phenyl]-2-propyl]acetamide

T2802-25G T2802-5G
TCT2802-5GEA 64.96 USD
TCT2802-5G TCT2802-25G
Thiamphenicol ≥98.0% (by HPLC, total nitrogen)
Thiamphenicol

CAS Number: 15318-45-3
MDL Number: MFCD00467983
Molecular Formula: C12H15Cl2NO5S
Molecular Weight: 356.21
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 166
Specific rotation [a]20/D: 11 deg (C=1, EtOH)


Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: 2,2-Dichloro-N-[(1R,2R)-1,3-dihydroxy-1-[4-(methylsulfonyl)phenyl]-2-propyl]acetamide

Formula: C₁₂H₁₅Cl₂NO₅S
MW: 356.21 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00467983
CAS Number: 15318-45-3
https://us.vwr.com/stibo/web/std.lang.all/64/78/28996478.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR