D-Threonine benzyl ester hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: H-D-Thr-OBzl.HCl

T2788-5G T2788-1G
TCT2788-5GEA 224 USD
TCT2788-5G TCT2788-1G
D-Threonine benzyl ester hydrochloride ≥98.0% (by HPLC, total nitrogen)
D-Threonine benzyl ester hydrochloride

CAS Number: 75748-36-6
Molecular Formula: C11H15NO3
Molecular Weight: 245.70
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 130
Specific rotation [a]20/D: 11 deg (C=1, EtOH)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Thr-OBzl.HCl

Formula: C₁₁H₁₅NO₃
MW: 245.70 g/mol
Melting Pt: 130 °C
MDL Number: MFCD28975347
CAS Number: 75748-36-6
https://us.vwr.com/stibo/web/std.lang.all/64/68/28996468.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR