3-(Trifluoromethyl)pyridine ≥98.0% (by GC)

Supplier: TCI America

T2786-25G T2786-5G
TCT2786-5GEA 70 USD
TCT2786-5G TCT2786-25G
3-(Trifluoromethyl)pyridine ≥98.0% (by GC)
3-(Trifluoromethyl)pyridine

CAS Number: 3796-23-4
MDL Number: MFCD00100032
Molecular Formula: C6H4F3N
Molecular Weight: 147.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 113
Flash Point (°C): 23
Specific Gravity (20/20): 1.28

Formula: C₆H₄F₃N
Boiling Pt: 113 °C
Density: 1.28 (20 °C)
Flash Pt: 23 °C
MDL Number: MFCD00100032
CAS Number: 3796-23-4
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR