2,3,5,6-Tetrabromothieno[3,2-b]thiophene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Perbromothieno[3,2-b]thiophene

T2772-1G
TCT2772-1GEA 282 USD
TCT2772-1G
2,3,5,6-Tetrabromothieno[3,2-b]thiophene ≥98.0% (by GC)
2,3,5,6-Tetrabromothieno[3,2-b]thiophene

CAS Number: 124638-53-5
MDL Number: MFCD03788229
Molecular Formula: C6Br4S2
Molecular Weight: 455.80
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 234

Formula: C₆Br₄S₂
MW: 455.80 g/mol
Melting Pt: 234 °C
MDL Number: MFCD03788229
CAS Number: 124638-53-5
https://us.vwr.com/stibo/web/std.lang.all/64/56/28996456.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR