α'-Bromo-α,α,α-trifluoro-o-xylene ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(Trifluoromethyl)benzyl bromide 2-(Bromomethyl)benzotrifluoride

T2768-25G T2768-5G
TCT2768-5GEA 17.26 USD
TCT2768-5G TCT2768-25G
α'-Bromo-α,α,α-trifluoro-o-xylene ≥97.0% (by GC, titration analysis)
α'-Bromo-α,α,α-trifluoro-o-xylene

CAS Number: 395-44-8
MDL Number: MFCD00013558
Molecular Formula: C8H6BrF3
Molecular Weight: 239.04
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 72
Flash Point (°C): 83
Freezing point (°C): 26


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(Bromomethyl)benzotrifluoride, alpha-Bromo-alpha',alpha',alpha'-trifluoro-o-xylene

Formula: C₈H₆BrF₃
MW: 239.03 g/mol
Boiling Pt: 72 °C
Flash Pt: 83 °C
MDL Number: MFCD00013558
CAS Number: 395-44-8
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/64/52/28996452.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR