2-(Trifluoromethyl)phenylhydrazinium chloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 1-(2-(Trifluoromethyl)phenyl)hydrazine hydrochloride 2-(Trifluoromethyl)phenylhydrazine hydrochloride 2-Hydrazinobenzotrifluoride hydrochloride

T2766-25G T2766-5G
TCT2766-25GEA 500.27 USD
TCT2766-25G TCT2766-5G
2-(Trifluoromethyl)phenylhydrazinium chloride ≥98.0% (by HPLC)
2-(Trifluoromethyl)phenylhydrazinium chloride

CAS Number: 3107-34-4
MDL Number: MFCD00102619
Molecular Formula: C7H7F3N2
Molecular Weight: 212.60
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 223


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Hydrazinobenzotrifluoride Hydrochloride

Formula: C₇H₈ClF₃N₂
MW: 212.6 g/mol
Melting Pt: 223 °C
MDL Number: MFCD00102619
CAS Number: 3107-34-4
https://us.vwr.com/stibo/web/std.lang.all/64/51/28996451.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR