4-Thiouracil ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4-Thioxo-3,4-dihydropyrimidin-2(1H)-one
CAS Number: 591-28-6
MDL Number: MFCD00090842
Molecular Formula: C4H4N2OS
Molecular Weight: 128.15
Purity/Analysis Method:
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 295
Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 2-Hydroxy-4-mercaptopyrimidine, 3,4-Dihydro-4-thioxo-2(1H)-pyrimidinone
Formula:
C₄H₄N₂OS MW: 128.15 g/mol Melting Pt: 295 °C |
MDL Number:
MFCD00090842 CAS Number: 591-28-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...