2-Bromo-4'-(trifluoromethyl)acetophenone ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
4-(Trifluoromethyl)phenacyl bromide
, 2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one
T2747-5G
T2747-25G
TCT2747-25GEA
297.4
USD
TCT2747-25G
TCT2747-5G
2-Bromo-4'-(trifluoromethyl)acetophenone ≥95.0% (by GC)
2-Bromo-4'-(trifluoromethyl)acetophenone
CAS Number: 383-53-9
MDL Number: MFCD00126489
Molecular Formula: C9H6BrF3O
Molecular Weight: 267.05
Purity/Analysis Method:
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 55
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 4-(Trifluoromethyl)phenacyl Bromide
Formula:
C₉H₆BrF₃O MW: 267.0400 g/mol Melting Pt: 55 °C |
MDL Number:
MFCD00126489 CAS Number: 383-53-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...