1-(4-(Trifluoromethyl)phenyl)piperazine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N-(α,α,α-Trifluoro-p-tolyl)piperazine
T2741-25G
T2741-5G
TCT2741-25GEA
174
USD
TCT2741-25G
TCT2741-5G
1-(4-(Trifluoromethyl)phenyl)piperazine ≥98.0% (by GC, titration analysis)
1-(4-(Trifluoromethyl)phenyl)piperazine
CAS Number: 30459-17-7
MDL Number: MFCD00040765
Molecular Formula: C11H13F3N2
Molecular Weight: 230.23
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 90
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-(1-Piperazinyl)benzotrifluoride
Formula:
C₁₁H₁₃F₃N₂ Melting Pt: 90 °C |
MDL Number:
MFCD00040765 CAS Number: 30459-17-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...