1-(2-(Trifluoromethyl)phenyl)piperazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T2739-1G T2739-5G
TCT2739-1GEA 160.44 USD
TCT2739-1G TCT2739-5G
1-(2-(Trifluoromethyl)phenyl)piperazine ≥98.0% (by GC, titration analysis)
1-(2-(Trifluoromethyl)phenyl)piperazine

CAS Number: 63854-31-9
MDL Number: MFCD02094727
Molecular Formula: C11H13F3N2
Molecular Weight: 230.23
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.23


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(1-Piperazinyl)benzotrifluoride

Formula: C₁₁H₁₃F₃N₂
Density: 1.23 (20 °C)
MDL Number: MFCD02094727
CAS Number: 63854-31-9
https://us.vwr.com/stibo/web/std.lang.all/64/27/28996427.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR