Nonyltrimethylammonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Nonyltrimethylammonium bromideTrimethylnonylammonium bromide

T2734-5G
TCT2734-5GEA 84.79 USD
TCT2734-5G
Nonyltrimethylammonium bromide ≥98.0% (by titrimetric analysis)
Nonyltrimethylammonium bromide

CAS Number: 1943-11-9
MDL Number: MFCD00041986
Molecular Formula: C12H28BrN
Molecular Weight: 266.27
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 223


Product Categories: Materials Chemistry, Cationic Surfactants, Bioscience, Quaternary Ammonium Salts for Clinical Chemistry
Other Product Names / Synonyms: Nonyltrimethylammonium Bromide

Formula: C₁₂H₂₈BrN
Melting Pt: 223 °C
MDL Number: MFCD00041986
CAS Number: 1943-11-9
https://us.vwr.com/stibo/web/std.lang.all/64/23/28996423.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR