2-(p-Toluenesulfonyloxy)acetic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-(p-Toluenesulfonyloxy)acetic acid
, 2-(p-Toluenesulphonyloxy)acetic acid
T2727-1G
TCT2727-1GEA
241.49
USD
TCT2727-1G
2-(p-Toluenesulfonyloxy)acetic acid ≥97.0% (by GC, titration analysis)
2-(p-Toluenesulfonyloxy)acetic acid
CAS Number: 39794-77-9
MDL Number: MFCD00021764
Molecular Formula: C9H10O5S
Molecular Weight: 230.23
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 140
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 2-(Tosyloxy)acetic Acid
Formula:
C₉H₁₀O₅S MW: 230.23 g/mol Melting Pt: 140 °C |
MDL Number:
MFCD00021764 CAS Number: 39794-77-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...