(3,3,3-Trifluoropropyl)trimethoxysilane ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Trimethoxy(3,3,3-trifluoropropyl)silane
T2720-25G
T2720-5G
TCT2720-5GEA
36.52
USD
TCT2720-5G
TCT2720-25G
(3,3,3-Trifluoropropyl)trimethoxysilane ≥98.0% (by GC)
(3,3,3-Trifluoropropyl)trimethoxysilane
CAS Number: 429-60-7
MDL Number: MFCD00039266
Molecular Formula: C6H13F3O3Si
Molecular Weight: 218.25
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 144
Flash Point (°C): 38
Specific Gravity (20/20): 1.14
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: 1,1,1-Trifluoro-3-(trimethoxysilyl)propane
Formula:
F₃CCH₂CH₂Si(OCH₃)₃ MW: 218.25 g/mol Boiling Pt: 144 °C Density: 1.14 (20 °C) Flash Pt: 38 °C |
MDL Number:
MFCD00039266 CAS Number: 429-60-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...