N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

Synonyms: N-Succinimidyl 2,2,2-Trichloroethyl CarbonateCarbonic Acid N-Succinimidyl 2,2,2-Trichloroethyl Ester

T2713-5G
TCT2713-5GEA 128 USD
TCT2713-5G
N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide ≥98.0% (by GC, total nitrogen)
N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide

CAS Number: 66065-85-8
MDL Number: MFCD00075216
Molecular Formula: C7H6Cl3NO5
Molecular Weight: 290.48
Purity/Analysis Method: 98.0% (GC,N)
Form: Crystal
Color: White
Melting point (°C): 113
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: N-Succinimidyl 2,2,2-Trichloroethyl Carbonate, Carbonic Acid N-Succinimidyl 2,2,2-Trichloroethyl Ester

Formula: C₇H₆Cl₃NO₅
MW: 290.48 g/mol
Melting Pt: 113 °C
MDL Number: MFCD00075216
CAS Number: 66065-85-8
https://us.vwr.com/stibo/web/std.lang.all/64/03/28996403.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR