Tetraisobutyl orthotitanate
Supplier: TCI America
Synonyms:
Isobutyl Titanate
T2711-5G
TCT2711-5GEA
151
USD
TCT2711-5G
Tetraisobutyl orthotitanate
Tetraisobutyl orthotitanate
CAS Number: 7425-80-1
MDL Number: MFCD00015045
Molecular Formula: C16H36O4Ti
Molecular Weight: 340.33
Form: Crystal
Color: White
Boiling point (°C): 141
Melting point (°C): 40
Other Product Names / Synonyms: Isobutyl Titanate, Titanium Isobutoxide
Formula:
C₁₆H₃₆O₄Ti MW: 340.33 g/mol Boiling Pt: 141 °C Melting Pt: 40 °C |
MDL Number:
MFCD00015045 CAS Number: 7425-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...