4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture) ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-(Trifluoromethyl)cyclohexanol
, 4-(Trifluoromethyl)cyclohexan-1-ol
T2706-5G
TCT2706-5GEA
240
USD
TCT2706-5G
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture) ≥98.0% (by GC)
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture)
CAS Number: 30129-18-1
MDL Number: MFCD00102144
Molecular Formula: C7H11F3O
Molecular Weight: 168.16
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 181
Specific Gravity (20/20): 1.23
Formula:
C₇H₁₁F₃O MW: 168.16 g/mol Boiling Pt: 181 °C Density: 1.23 (20 °C) |
MDL Number:
MFCD00102144 CAS Number: 30129-18-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...