Tetradecanal ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Tetradecylaldehyde

T2696-1G
TCT2696-1GEA 373.41 USD
TCT2696-1G
Tetradecanal ≥95.0% (by GC)
Tetradecanal

CAS Number: 124-25-4
MDL Number: MFCD00007019
Molecular Formula: C14H28O
Molecular Weight: 212.38
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 166
Melting point (°C): 23
Storage Temperature: 0°C


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Tetradecanaldehyde, Tetradecyl Aldehyde, Myristaldehyde, Myristyl Aldehyde

Formula: C₁₄H₂₈O
MW: 212.38 g/mol
Boiling Pt: 166 °C
Melting Pt: 23 °C
MDL Number: MFCD00007019
CAS Number: 124-25-4
https://us.vwr.com/stibo/web/std.lang.all/63/91/28996391.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR