(1R,4R,5R)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane ≥95.0% (by GC)

Supplier: TCI America

T2578-5G T2578-1G
TCT2578-1GEA 163.37 USD
TCT2578-1G TCT2578-5G
(1R,4R,5R)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane ≥95.0% (by GC)
(1R,4R,5R)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane

CAS Number: 5718-75-2
Molecular Formula: C10H18S
Molecular Weight: 170.31
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 84
Specific Gravity (20/20): 1.00
Specific rotation [a]20/D: -70 deg (C=1.32, CHCl3)

Formula: C₁₀H₁₈S
Boiling Pt: 84 °C
Density: 1.00 (20 °C)
CAS Number: 5718-75-2
https://us.vwr.com/stibo/web/std.lang.all/62/92/28996292.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR