4,4,5,5-Tetramethyl-1,3,2-dioxaborolane ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Pinacol Borane

T2572-25G T2572-5G
TCT2572-25GEA 159.04 USD
TCT2572-25G TCT2572-5G
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane ≥97.0% (by GC)
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane

CAS Number: 25015-63-8
MDL Number: MFCD00674030
Molecular Formula: C6H13BO2
Molecular Weight: 127.98
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 43
Flash Point (°C): 5
Specific Gravity (20/20): 0.89


Product Categories: Synthetic Organic Chemistry, Boration
Other Product Names / Synonyms: Pinacol Borane

Formula: C₆H₁₃BO₂
MW: 127.98 g/mol
Boiling Pt: 43 °C
Density: 0.89 (20 °C)
Flash Pt: 5 °C
MDL Number: MFCD00674030
CAS Number: 25015-63-8
https://us.vwr.com/stibo/web/std.lang.all/62/86/28996286.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR