Tetrabutylphosphonium hydroxide 40% in water
Supplier: TCI America
T2571-25G
T2571-250G
TCT2571-25GEA
35.51
USD
TCT2571-25G
TCT2571-250G
Tetrabutylphosphonium hydroxide 40% in water
Tetrabutylphosphonium hydroxide
CAS Number: 14518-69-5
MDL Number: MFCD00068456
Molecular Formula: C16H37OP
Molecular Weight: 276.44
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: TBPH
Formula:
C₁₆H₃₇OP Density: 0.99 (20 °C) |
MDL Number:
MFCD00068456 CAS Number: 14518-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...