α'-Chloro-α,α,α-trifluoro-p-xylene ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
1-(Chloromethyl)-4-(trifluoromethyl)benzene
CAS Number: 939-99-1
MDL Number: MFCD00040772
Molecular Formula: C8H6ClF3
Molecular Weight: 194.58
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 68
Melting point (°C): 19
Flash Point (°C): 74
Specific Gravity (20/20): 1.32
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-(Chloromethyl)benzotrifluoride, alpha-Chloro-alpha',alpha',alpha'-trifluoro-p-xylene
Formula:
C₈H₆ClF₃ MW: 194.58 g/mol Boiling Pt: 68 °C Melting Pt: 19 °C Density: 1.32 (20 °C) Flash Pt: 74 °C |
MDL Number:
MFCD00040772 CAS Number: 939-99-1 UN: 3265 ADR: 8,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...