TCIAT2472

T2472-5G
TCT2472-5GEA 84.4 USD
TCT2472-5G
TCIAT2472
CAS Number: 4913-13-7
MDL Number: MFCD00008307
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Color: Yellow
Boiling point (°C): 224
Flash Point (°C): 90
Specific Gravity (20/20): 0.93

Other Product Names / Synonyms: 5-(Dimethylamino)-m-xylene, N,N-Dimethyl-3,5-xylidine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR