Tetrathiafulvalene-7,7,8,8-tetracyanoquinodimethane complex ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: TTF - TCNQ Complex

T2468-1G
TCT2468-1GEA 298.41 USD
TCT2468-1G
Tetrathiafulvalene-7,7,8,8-tetracyanoquinodimethane complex ≥98.0% (by total nitrogen basis)
Tetrathiafulvalene-7,7,8,8-tetracyanoquinodimethane complex

CAS Number: 40210-84-2
MDL Number: MFCD03791115
Molecular Formula: C6H4S4
Molecular Weight: 408.53
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Color: Purple
Melting point (°C): 226


Product Categories: Materials Chemistry, Molecular Conductors, Molecular Conductors
Other Product Names / Synonyms: TTF - TCNQ Complex

Formula: C₆H₄S₄
MW: 408.53 g/mol
Melting Pt: 226 °C
MDL Number: MFCD03791115
CAS Number: 40210-84-2
https://us.vwr.com/stibo/web/std.lang.all/62/06/28996206.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR