1,3,5-Tris[4-(trifluoromethanesulfonyloxy)-3-(trimethylsilyl)phenyl]benzene ≥92.0% (by HPLC)

Supplier: TCI America

T2467-1G
TCT2467-1GEA 574 USD
TCT2467-1G
1,3,5-Tris[4-(trifluoromethanesulfonyloxy)-3-(trimethylsilyl)phenyl]benzene ≥92.0% (by HPLC)
1,3,5-Tris[4-(trifluoromethanesulfonyloxy)-3-(trimethylsilyl)phenyl]benzene

CAS Number: 847925-63-7
Molecular Formula: C36H39F9O9S3Si3
Molecular Weight: 967.12
Purity/Analysis Method: 92.0% (HPLC)
Form: Crystal
Color: White

Formula: C₃₆H₃₉F₉O₉S₃Si₃
CAS Number: 847925-63-7
https://us.vwr.com/stibo/web/std.lang.all/62/05/28996205.jpg

Order Now

Specification Test Results

Purity/Analysis Method 92.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR