2-(Trifluoromethoxy)benzonitrile ≥98.0% (by GC)

Supplier: TCI America

T2371-5G
TCT2371-5GEA 143 USD
TCT2371-5G
2-(Trifluoromethoxy)benzonitrile ≥98.0% (by GC)
2-(Trifluoromethoxy)benzonitrile

CAS Number: 63968-85-4
MDL Number: MFCD00042409
Molecular Formula: C8H4F3NO
Molecular Weight: 187.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 62
Specific Gravity (20/20): 1.31

Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Boiling Pt: 62 °C
Density: 1.31 (20 °C)
MDL Number: MFCD00042409
CAS Number: 63968-85-4
UN: 3276
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/61/38/28996138.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR