4-(Trifluoromethyl)phenylacetonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-(Trifluoromethyl)benzyl cyanide

T2368-5G
TCT2368-5GEA 30.45 USD
TCT2368-5G
4-(Trifluoromethyl)phenylacetonitrile ≥98.0% (by GC)
4-(Trifluoromethyl)phenylacetonitrile

CAS Number: 2338-75-2
MDL Number: MFCD00001921
Molecular Formula: C9H6F3N
Molecular Weight: 185.15
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 132
Melting point (°C): 47
Flash Point (°C): 113


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-(Trifluoromethyl)benzyl Cyanide

Formula: C₉H₆F₃N
MW: 185.15 g/mol
Boiling Pt: 132 °C
Melting Pt: 47 °C
Flash Pt: 113 °C
MDL Number: MFCD00001921
CAS Number: 2338-75-2
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/61/35/28996135.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR