TCIAT2277

T2277-1G
TCT2277-1GEA 210.4 USD
TCT2277-1G
TCIAT2277
CAS Number: 1378304-83-6
Molecular Formula: C6H10F3N
Molecular Weight: 153.15
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.11
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR