Tetrakis(dimethylamino)titanium (TDMAT) ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Tetrakis(dimethylamino)titanium(IV)
, TDMAT
T2230-1G
T2230-5G
TCT2230-1GEA
48.72
USD
TCT2230-1G
TCT2230-5G
Tetrakis(dimethylamino)titanium (TDMAT) ≥97.0% (by titrimetric analysis)
Tetrakis(dimethylamino)titanium (TDMAT)
CAS Number: 3275-24-9
Molecular Formula: C8H24N4Ti
Molecular Weight: 224.18
Purity/Analysis Method:
Form: Clear Liquid
Color: Pale Yellow
Flash Point (°C): -30
Specific Gravity (20/20): 0.95
Formula:
C₈H₂₄N₄Ti MW: 224.17 g/mol Density: 0.95 (20 °C) Flash Pt: –30 °C |
MDL Number:
MFCD00014861 UN: 3129 ADR: 4.3,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...