1-Chloro-4-(trifluoromethoxy)benzene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-(Trifluoromethoxy)chlorobenzene
T2149-5G
TCT2149-5GEA
34.48
USD
TCT2149-5G
1-Chloro-4-(trifluoromethoxy)benzene ≥98.0% (by GC)
1-Chloro-4-(trifluoromethoxy)benzene
CAS Number: 461-81-4
MDL Number: MFCD00040953
Molecular Formula: C7H4ClF3O
Molecular Weight: 196.55
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 145
Specific Gravity (20/20): 1.37
Formula:
C₇H₄ClF₃O MW: 196.56 g/mol Boiling Pt: 145 °C Density: 1.37 (20 °C) |
MDL Number:
MFCD00040953 CAS Number: 461-81-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...