Dimethylthexylchlorosilane ≥95.0% (by GC)

Supplier: TCI America

T2116-25G T2116-5G
TCT2116-5GEA 108 USD
TCT2116-5G TCT2116-25G
Dimethylthexylchlorosilane ≥95.0% (by GC)
Dimethylthexylchlorosilane

CAS Number: 67373-56-2
MDL Number: MFCD00012175
Molecular Formula: C8H19ClSi
Molecular Weight: 178.78
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 56
Flash Point (°C): 46
Specific Gravity (20/20): 0.91


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis), Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Chlorodimethyl(1,1,2-trimethylpropyl)silane, Dimethylthexylsilyl Chloride

Formula: C₈H₁₉ClSi
Boiling Pt: 56 °C
Density: 0.91 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00012175
CAS Number: 67373-56-2
UN: 2986
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/60/01/28996001.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR