2,4,6,8-Tetramethylcyclotetrasiloxane ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
1,3,5,7-Tetramethylcyclotetrasiloxane
T2076-5G
T2076-25G
TCT2076-5GEA
31.47
USD
TCT2076-5G
TCT2076-25G
2,4,6,8-Tetramethylcyclotetrasiloxane ≥95.0% (by GC)
2,4,6,8-Tetramethylcyclotetrasiloxane
CAS Number: 2370-88-9
MDL Number: MFCD00039567
Molecular Formula: C4H16O4Si4
Molecular Weight: 240.51
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 134
Flash Point (°C): 24
Specific Gravity (20/20): 0.99
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1,3,5,7-Tetramethylcyclotetrasiloxane, TMCTS
Formula:
C₄H₁₆O₄Si₄ MW: 240.51 g/mol Boiling Pt: 134 °C Density: 0.99 (20 °C) Flash Pt: 24 °C |
MDL Number:
MFCD00039567 CAS Number: 2370-88-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...