Tetrabutylphosphonium tetrafluoroborate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

T2006-25G
TCT2006-25GEA 132 USD
TCT2006-25G
Tetrabutylphosphonium tetrafluoroborate ≥97.0% (by titrimetric analysis)
Tetrabutylphosphonium tetrafluoroborate

CAS Number: 1813-60-1
MDL Number: MFCD00270144
Molecular Formula: C16H36BF4P
Molecular Weight: 346.24
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: White

Formula: C₁₆H₃₆BF₄P
MW: 346.2354 g/mol
MDL Number: MFCD00270144
CAS Number: 1813-60-1
https://us.vwr.com/stibo/web/std.lang.all/59/47/28995947.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR