1,5,7-Triazabicyclo[4.4.0]dec-5-ene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Hhpp
, 2,3,4,6,7,8-Hexahydro-1H-pyrimido[1,2-a]pyrimidine
, TBD
, 1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine
T1982-5G
TCT1982-5GEA
166.01
USD
TCT1982-5G
1,5,7-Triazabicyclo[4.4.0]dec-5-ene ≥98.0% (by HPLC, titration analysis)
1,5,7-Triazabicyclo[4.4.0]dec-5-ene
CAS Number: 5807-14-7
MDL Number: MFCD00043003
Molecular Formula: C7H13N3
Molecular Weight: 139.20
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 130
Other Product Names / Synonyms: 1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, TBD
Formula:
C₇H₁₃N₃ MW: 139.20 g/mol Melting Pt: 130 °C |
MDL Number:
MFCD00043003 CAS Number: 5807-14-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...