1,2,5-Trimethylpyrrole ≥98.0% (by GC)
Supplier: TCI America
T1939-5ML
T1939-25ML
TCT1939-25MLEA
144.13
USD
TCT1939-25ML
TCT1939-5ML
1,2,5-Trimethylpyrrole ≥98.0% (by GC)
1,2,5-Trimethylpyrrole
CAS Number: 930-87-0
MDL Number: MFCD00003090
Molecular Formula: C7H11N
Molecular Weight: 109.17
Purity/Analysis Method:
Form: Clear Liquid
Color: Very Pale Yellow
Flash Point (°C): 52
Specific Gravity (20/20): 0.92
Formula:
C₇H₁₁N MW: 109.1689 g/mol Density: 0.92 (20 °C) Flash Pt: 52 °C |
MDL Number:
MFCD00003090 CAS Number: 930-87-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...