(2,4,6-Trimethylphenyl)acetonitrile ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Mesitylacetonitrile
, 2,4,6-Trimethylbenzyl cyanide
, Mesitylacetonitrile
T1938-5G
TCT1938-5GEA
68.01
USD
TCT1938-5G
(2,4,6-Trimethylphenyl)acetonitrile ≥98.0% (by GC)
(2,4,6-Trimethylphenyl)acetonitrile
CAS Number: 34688-71-6
MDL Number: MFCD00013815
Molecular Formula: C11H13N
Molecular Weight: 159.23
Purity/Analysis Method:
Form: Crystal
Color: White
Other Product Names / Synonyms: Mesitylacetonitrile, 2,4,6-Trimethylphenylacetonitrile
Formula:
C₁₁H₁₃N MW: 159.23 g/mol |
MDL Number:
MFCD00013815 CAS Number: 34688-71-6 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...