2,3,4,5-Tetrafluorobenzaldehyde ≥96.0% (by GC)

Supplier: TCI America

T1897-5G T1897-1G
TCT1897-5GEA 446 USD
TCT1897-5G TCT1897-1G
2,3,4,5-Tetrafluorobenzaldehyde ≥96.0% (by GC)
2,3,4,5-Tetrafluorobenzaldehyde

CAS Number: 16583-06-5
MDL Number: MFCD02683041
Molecular Formula: C7H2F4O
Molecular Weight: 178.09
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 82
Specific Gravity (20/20): 1.51

Formula: C₇H₂F₄O
MW: 178.09 g/mol
Boiling Pt: 82 °C
Density: 1.51 (20 °C)
MDL Number: MFCD02683041
CAS Number: 16583-06-5
https://us.vwr.com/stibo/web/std.lang.all/58/58/28995858.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR