Triphenylantimony dichloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Triphenylantimony(V) dichloride
T1849-1G
TCT1849-1GEA
83.23
USD
TCT1849-1G
Triphenylantimony dichloride ≥98.0% (by titrimetric analysis)
Triphenylantimony dichloride
CAS Number: 594-31-0
MDL Number: MFCD00013598
Molecular Formula: C18H15Cl2Sb
Molecular Weight: 423.98
Purity/Analysis Method:
Form: Crystal
Color: White
Other Product Names / Synonyms: Dichlorotriphenylantimony, Triphenylstibine Dichloride
Formula:
C₁₈H₁₅Cl₂Sb MW: 423.98 g/mol |
MDL Number:
MFCD00013598 CAS Number: 594-31-0 UN: 2923 ADR: 8,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...