4-(Trifluoromethoxy)anisole ≥96.0% (by GC)

Supplier: TCI America

Synonyms: 1-Methoxy-4-(trifluoromethoxy)benzene

T1779-5G
TCT1779-5GEA 65 USD
TCT1779-5G
4-(Trifluoromethoxy)anisole ≥96.0% (by GC)
4-(Trifluoromethoxy)anisole

CAS Number: 710-18-9
MDL Number: MFCD00216942
Molecular Formula: C8H7F3O2
Molecular Weight: 192.14
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 58
Specific Gravity (20/20): 1.28

Formula: C₈H₇F₃O₂
MW: 192.14 g/mol
Density: 1.28 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00216942
CAS Number: 710-18-9
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/57/54/28995754.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR