2,3,5,6-Tetrafluoropyridine ≥96.0% (by GC)

Supplier: TCI America

T1778-1G T1778-5G
TCT1778-5GEA 146.16 USD
TCT1778-5G TCT1778-1G
2,3,5,6-Tetrafluoropyridine ≥96.0% (by GC)
2,3,5,6-Tetrafluoropyridine

CAS Number: 2875-18-5
MDL Number: MFCD00011734
Molecular Formula: C5HF4N
Molecular Weight: 151.06
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 101
Flash Point (°C): 31
Specific Gravity (20/20): 1.52

Formula: C₅HF₄N
MW: 151.06 g/mol
Boiling Pt: 101 °C
Density: 1.52 (20 °C)
Flash Pt: 31 °C
MDL Number: MFCD00011734
CAS Number: 2875-18-5
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/57/53/28995753.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR