Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Trifluoromethoxy)benzene ≥98.0% (by GC)

Supplier: TCI America


T1617-25G
TCT1617-25GEA 56.19 USD InStock
TCT1617-25G
(Trifluoromethoxy)benzene ≥98.0% (by GC)
(Trifluoromethoxy)benzene
CAS Number: 456-55-3
MDL Number: MFCD00040832
Molecular Formula: C7H5F3O
Molecular Weight: 162.11
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 104
Flash Point (°C): 12
Specific Gravity (20/20): 1.23

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoroanisole


Formula: C₇H₅F₃O
MW: 162,11 g/mol
Boiling Pt: 104 °C
Melting Pt: -50 °C
Density: 1,23 (20 °C)
Flash Pt: 12 °C
MDL Number: MFCD00040832
CAS Number: 456-55-3
EINECS: 207-269-5
UN: 1993
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/72/25/757225.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty