(2S)-(+)-2,3-Epoxypropyl toluene-4-sulfonate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(2S)-(+)-Glycidyl tosylate
, (s)-(+)-oxirane-2-methanol p-toluenesulfonate
, (2S)-(+)-Glycidyl p-toluenesulfonate
, (2S)-(+)-Glycidyl p-toluenesulphonate
, (s)-(+)-oxirane-2-methanol p-toluenesulphonate
CAS Number: 70987-78-9
MDL Number: MFCD00064489
Molecular Formula: C10H12O4S
Molecular Weight: 228.26
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 48
Flash Point (°C): 202
Specific rotation [a]20/D: 19 deg (C=2.5, CH3CN)
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Chiral Building Blocks
Other Product Names / Synonyms: (2S)-(+)-Glycidyl Tosylate, p-Toluenesulfonic Acid (2S)-(+)-Glycidyl Ester
Formula:
C₁₀H₁₂O₄S MW: 228.26 g/mol Melting Pt: 48 °C Flash Pt: 202 °C |
MDL Number:
MFCD00064489 CAS Number: 70987-78-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...